Anti-CA9 Mouse Monoclonal Antibody (Purified) [clone: PN-15]

Lieferant: Biotium
BNUB0637-500 BNUB0637-100
BNUB0637-500EA 511 EUR
BNUB0637-500 BNUB0637-100
Anti-CA9 Mouse Monoclonal Antibody (Purified) [clone: PN-15]
Antikörper
Recognizes a glycoprotein of ~200 kDa, identified as carbonic anhydrase IX (CAIX/gp200). Its epitope resides in the carbohydrate domain of gp200. It shows no significant cross-reactivity with other carbohydrate determinants, such as the Lewis blood group antigens, epithelial membrane antigen, HMFG, and AB blood group antigens. In normal kidney, gp200 is localized along the brush border of the pars convoluta and pars recta segments of the proximal tubule, as well as focally along the luminal surface of Bowman's capsule adjoining the outgoing proximal tubule. Reportedly, gp200 is expressed by 93% of primary and 84% of metastatic renal cell carcinomas. This MAb may be useful in the investigations of carcinomas of proximal nephrogenic differentiation especially those showing tubular differentiation.

Type: Primary
Antigen: CA9
Clonality: Monoclonal
Clone: PN-15
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Horse, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR