Anti-TGFA Mouse Monoclonal Antibody (Biotin) [clone: MF9]

Lieferant: Biotium
BNCB0644-500 BNCB0644-100
BNCB0644-500EA 560 EUR
BNCB0644-500 BNCB0644-100
Anti-TGFA Mouse Monoclonal Antibody (Biotin) [clone: MF9]
Antikörper
This antibody reacts with the C-terminus of TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. The staining with Ab-1 is completely blocked by the peptide used for raising this antibody. TGF alpha (aa50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.

Type: Primary
Antigen: TGFA
Clonality: Monoclonal
Clone: MF9
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Cow, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR