Anti-L1CAM Mouse Monoclonal Antibody (CF647) [clone: UJ127]
Lieferant: Biotium
BNC470679-500 BNC470679-100
BNC470679-500EA 560 EUR
BNC470679-500 BNC470679-100
Anti-L1CAM Mouse Monoclonal Antibody (CF647) [clone: UJ127]
Antikörper
Recognizes a cell surface protein of 220-240 kDa, identified as L1 cell adhesion molecule. The L1CAM gene, which is located in Xq28, is involved in three distinct conditions: 1) HSAS (hydrocephalus-stenosis of the aqueduct of Sylvius); 2) MASA (mental retardation, aphasia, shuffling gait, and adducted thumbs); and 3) SPG1 (spastic paraplegia). The L1, neural cell adhesion molecule (L1CAM) also plays an important role in axon growth, fasciculation, and neural migration as well as in mediating neuronal differentiation. Expression of L1 protein is restricted to tissues arising fromMore Product Information