Anti-MLANA Mouse Monoclonal Antibody [clone: HMB45 A103 T311]

Lieferant: Biotium
BNUB0702-500 BNUB0702-100
BNUB0702-500EA 511 EUR
BNUB0702-500 BNUB0702-100
Anti-MLANA Mouse Monoclonal Antibody [clone: HMB45 A103 T311]
Antikörper
This antibody cocktail recognizes three melanoma-specific proteins, which include MART-1, Tyrosinase and gp100. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Tyrosinase is one of the targets for cytotoxic T-cell recognition in melanoma patients. The function of gp100 is not known but it is reported to be a useful marker for melanocytes and melanomas. This cocktail of three markers is designed for extremely sensitive labeling of formalin-fixed, paraffin-embedded melanomas and other tumors showing melanocytic differentiation.

  • Purified

Type: Primary
Antigen: MLANA
Clonality: Monoclonal
Clone: HMB45 A103 T311
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR