Anti-ALPP Mouse Monoclonal Antibody (Biotin) [clone: ALPP/870]

Lieferant: Biotium
BNCB0870-500 BNCB0870-100
BNCB0870-500EA 560 EUR
BNCB0870-500 BNCB0870-100
Anti-ALPP Mouse Monoclonal Antibody (Biotin) [clone: ALPP/870]
Antikörper
Reacts with a 70 kDa membrane-bound isozyme (Regan and Nagao type) of Placental Alkaline Phosphatase (PLAP) occurring in the placenta during the 3rd trimester of gestation. It is highly specific for PLAP and shows no cross-reaction with other isozymes of alkaline phosphatase. Anti-PLAP reacts with germ cell tumors and can discriminate between these and other neoplasms. Somatic neoplasms e.g. breast, gastrointestinal, prostatic, and urinary cancers may also immunoreact with antibodies to PLAP. Anti-PLAP positivity in conjunction with anti-keratin negativity favors seminoma over carcinoma.More Product Information
Bestellen

TECHNISCHE DATEN

Antigen-Symbol ALPP
Environmentally Preferable
Antigen-Name PLAP
Antikörper-Typ Primär
Klonalität Monoklonal
Konjugation Biotin
Klon Clone ALPP/870
Reaktivität Human
Wirt Mouse
Gen-ID 284255
Isotyp IgG2b kappa
ImmunoChemie Yes
ImmunFluoreszenz Yes
Konzentration 0,1 mg/ml
Lagerungspuffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR