Anti-KRT8 & KRT18 Mouse Monoclonal Antibody (Biotin) [clone: KRT8/803 KRT18/835]

Lieferant: Biotium
BNCB0979-500 BNCB0979-100
BNCB0979-500EA 560 EUR
BNCB0979-500 BNCB0979-100
Anti-KRT8 & KRT18 Mouse Monoclonal Antibody (Biotin) [clone: KRT8/803 KRT18/835]
Antikörper
Cytokeratin 8 (CK8) belongs to the type II (or B or basic) subfamily of high molecular weight cytokeratins and exists in combination with cytokeratin 18 (CK18). This MAb cocktail recognizes all simple epithelia including glandular epithelium, for example thyroid, female breast, gastrointestinal tract, respiratory tract, and urogenital tract including transitional epithelium. All adenocarcinomas and most squamous carcinomas are positive but keratinizing squamous carcinomas are usually negative. This antibody is useful in demonstrating the presence of Paget cells; there is very little keratin 18 in the normal epidermis so only Paget cells are stained.Immuno-histochemical staining with this MAb is indistinguishable from that obtained with monoclonal antibody 5D3.

Type: Primary
Antigen: KRT8 & KRT18
Clonality: Monoclonal
Clone: KRT8/803 KRT18/835
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR