Anti-KIT Mouse Monoclonal Antibody (Biotin) [clone: Cocktail]

Lieferant: Biotium
BNCB1018-500 BNCB1018-100
BNCB1018-500EA 560 EUR
BNCB1018-500 BNCB1018-100
Anti-KIT Mouse Monoclonal Antibody (Biotin) [clone: Cocktail]
Antikörper
This MAb recognizes a protein of 145 kDa, identified as CD117/p145kit. It is found on a wide variety of tumor cells including follicular and papillary carcinoma of thyroid, adenocarcinomas from endometrium, lung, ovary, pancreas, and breast as well as malignant melanoma, endodermal sinus tumor, and small cell carcinoma. However, anti-CD117 has been particularly useful in differentiating gastrointestinal stromal tumors from Kaposi's sarcoma, tumors of smooth muscle origin, fibromatosis, and neural tumors of the GI tract. Anti-CD117 is also useful in recognizing myeloblasts in bone marrow biopsy and clot section.

Type: Primary
Antigen: KIT
Clonality: Monoclonal
Clone: Cocktail
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR