Anti-B2M Mouse Monoclonal Antibody (CF568) [clone: 246-E9.E7]

Lieferant: Biotium
BNC681039-500 BNC681039-100
BNC681039-500EA 560 EUR
BNC681039-500 BNC681039-100
Anti-B2M Mouse Monoclonal Antibody (CF568) [clone: 246-E9.E7]
Antikörper
Recognizes a protein of 12 kDa, identified as β-microglobulin. β-microglobulin non-covalently associates with the 44 kDa chain to form the HLA Class I antigen complex. Human β-2 microglobulin associated with HLA Class I antigens is expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells. It is absent on erythrocytes. This MAb is specific to human β-2 microglobulin and does not react with non-human primate cells. This antibody reacts with all cell types excluding erythrocytes. The detection of β-2 microglobulin in body fluids has been used as a tumor marker and for monitoring patients with HIV infection.

Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: 246-E9.E7
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR