Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF568) [clone: HLA-DRB/1067]
Lieferant: Biotium
BNC681067-500 BNC681067-100
BNC681067-500EA 560 EUR
BNC681067-500 BNC681067-100
Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF568) [clone: HLA-DRB/1067]
Antikörper
This MAb reacts with the beta-chain of HLA-DRB1 antigen, a member of MHC class II molecules. It does not cross react with HLA-DP and HLA-DQ. Its epitope is different from that of MAb L243. HLA-DR is a heterodimeric cell surface glycoprotein comprised of a 36 kDa alpha (heavy) chain and a 28 kDa beta (light) chain. It is expressed on B-cells, activated T-cells, monocytes/macrophages, dendritic cells and other non-professional APCs. In conjunction with the CD3/TCR complex and CD4 molecules, HLA-DR is critical for efficient peptide presentation to CD4 T cells. It is an excellent histiocyticMore Product Information