Anti-KRT77 Mouse Monoclonal Antibody (CF647) [clone: KRTL/1077]

Lieferant: Biotium
BNC471077-500 BNC471077-100
BNC471077-500EA 560 EUR
BNC471077-500 BNC471077-100
Anti-KRT77 Mouse Monoclonal Antibody (CF647) [clone: KRTL/1077]
Antikörper
This MAb recognizes the 56.5 kDa (CK10); 50 kDa (CK14); 50 kDa (CK15); 48 kDa (CK16); 40 kDa (CK19) keratins of the acidic (Type I or LMW) subfamily. Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. The acidic keratins have molecular weights (MW) of 56.5, 55, 51, 50, 50 , 48, 46, 45, and 40 kDa. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

Type: Primary
Antigen: KRT77
Clonality: Monoclonal
Clone: KRTL/1077
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR