Anti-HLA-DRA Mouse Monoclonal Antibody (CF594) [clone: 169-1B5.2]

Lieferant: Biotium
BNC941096-500 BNC401096-100
BNC941096-500EA 560 EUR
BNC941096-500 BNC401096-100
Anti-HLA-DRA Mouse Monoclonal Antibody (CF594) [clone: 169-1B5.2]
Antikörper
This antibody detects a monomorphic general framework determinant of HLA-DR Class II antigen. It does not cross react with HLA-DP and HLA-DQ. HLA-DR is a heterodimeric cell surface glycoprotein comprised of a 36 kDa alpha (heavy) chain and a 28 kDa beta (light) chain. It is expressed on B-cells, activated T-cells, monocytes/macrophages, dendritic cells and other non-professional APCs. In conjunction with the CD3/TCR complex and CD4 molecules, HLA-DR is critical for efficient peptide presentation to CD4 T cells. It is an excellent histiocytic marker in paraffin sections producing intense cytoplasmic staining. True histiocytic neoplasms are similarly positive. HLA-DR antigens also occur on a variety of epithelial cells and their corresponding neoplastic counterparts.

Type: Primary
Antigen: HLA-DRA
Clonality: Monoclonal
Clone: 169-1B5.2
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Cat, Cow, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR