Anti-HLA-DPB1 & HLA-DRA Mouse Monoclonal Antibody (Purified) [clone: Bra-14]

Lieferant: Biotium
BNUB1129-500 BNUB1129-100
BNUB1129-500EA 511 EUR
BNUB1129-500 BNUB1129-100
Anti-HLA-DPB1 & HLA-DRA Mouse Monoclonal Antibody (Purified) [clone: Bra-14]
Antikörper
Reacts with a common epitope of human major histocompatibility (MHC) class II antigens, HLA-DR and DP. Human MHC class II antigens are transmembrane glycoproteins composed of an chain (36 kDa) and a chain (27 kDa). They are expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, and thymic epithelial cells and are also present on activated T lymphocytes. Human MHC class II genes are located in the HLA-D region that encodes at least six and ten chain genes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides and present them to T-helper cells. They, therefore, have a critical role in the initiation of the immune response. It has been shown that some autoimmune diseases are associated with certain class II alleles.

Type: Primary
Antigen: HLA-DPB1 & HLA-DRA
Clonality: Monoclonal
Clone: Bra-14
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR