Anti-EPX Mouse Monoclonal Antibody (CF568) [clone: AHE-1]
Lieferant: Biotium
BNC681231-500 BNC681231-100
BTIUBNC681231-500EA 560 EUR
BTIUBNC681231-500 BTIUBNC681231-100
Anti-EPX Mouse Monoclonal Antibody (CF568) [clone: AHE-1]
Antikörper
Peripheral blood granulocytes are classified into neutrophils, basophils and eosinophils according to the staining characteristics of their cytoplasmic granules. Granule proteins are released by physiologic and pharmacologic stimuli and play important roles in both normal and pathological host immune responses. Eosinophil major basic protein and eosinophil peroxidase (EPX) are granule proteins specific to the eosinophil. AHE-1 recognizes human EPX, a granule protein specific to eosinophils. It does not cross-react with eosinophil major basic protein, elastase, cathepsin G, esterase N,More Product Information