Anti-PTH Mouse Monoclonal Antibody (CF640R) [clone: 3H9 PTH/1175]

Lieferant: Biotium
BNC401236-500 BNC401236-100
BNC401236-500EA 560 EUR
BNC401236-500 BNC401236-100
Anti-PTH Mouse Monoclonal Antibody (CF640R) [clone: 3H9 PTH/1175]
Antikörper
The epitope of this MAb maps in the C-terminus of PTH, a hormone produced by the parathyroid gland that regulates the concentration of calcium and phosphorus in extracellular fluid. This hormone elevates blood Ca2 levels by dissolving the salts in bone and preventing their renal excretion.It is produced in the parathyroid gland as an 84 amino acid single chain polypeptide. It can also be secreted as N-terminal truncated fragments or C-terminal fragments after intracellular degradation, as in case of hypercalcemia. Defects in this gene are a cause of familial isolated hypoparathyroidism (FIH); also called autosomal dominant hypoparathyroidism or autosomal dominant hypocalcemia. FIH is characterized by hypocalcemia and hyperphosphatemia due to inadequate secretion of parathyroid hormone. Symptoms are seizures, tetany and cramps. FIH exist both as autosomal dominant and recessive forms of hypoparathyroidism.

Type: Primary
Antigen: PTH
Clonality: Monoclonal
Clone: 3H9 PTH/1175
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR