Gas sampling bags, ALTEF®
Lieferant: Restek
REST22962EA 556 EUR
REST22962 REST26329 REST22961 REST22963 REST26328 REST22959 REST26330 REST26331 REST22960
Gas sampling bags, ALTEF®
Beutel Gasbeutel
Gas sampling bags are whole air sampling devices useful for monitoring part-per million (ppm) levels of most volatile organic compounds (VOCs). Unlike Tedlar® bags, ALTEF bags do not produce background levels of dimethylacetamide (DMAC) or phenol. It features single eyelet and PP combo valve with hose connection to fit 4,7 mm ID tubing and syringe port with replaceable septum.
  • Durable polyvinylidene fluoride (PVDF) film is resistant to abrasion and chemicals, including most acids and organic compounds
  • Excellent alternative to Tedlar® bags for collection of most VOCs
  • Very low VOC and sulfur background compared to Tedlar® bags
  • Not recommended for ketones, acetates, hydrogen sulfide or permanent gases
  • Contain no additives, fillers, or pigments
  • 82 °C (180 °F) maximum operating temperature
Note: Use only Point Style 5 syringes.