Ac-WEHD-AFC
Lieferant: ENZO LIFE SCIENCES
ALX260117M005 ALX260117M010
ENZOALX260117M005EA 316 EUR
ENZOALX260117M005 ENZOALX260117M010
Ac-WEHD-AFC
Proteine und Peptide
Substrate for caspase-1, -4- and -5
Fluorogenic substrate for caspase-1 and -5. Also a substrate for caspase-4.