Ac-YVAD-CHO
Lieferant: ENZO LIFE SCIENCES
ALX260027M001 ALX260027M005
ENZOALX260027M001EA 90.2 EUR
ENZOALX260027M001 ENZOALX260027M005
Ac-YVAD-CHO
Proteine und Peptide
Inhibitor of caspase-1 and -4
Potent and reversible inhibitor of caspase-1. Also inhibits caspase-4. Strongly inhibits anti-Fas induced apoptosis in L929-Fas cells.