Z-WEHD-FMK
Lieferant: ENZO LIFE SCIENCES
ALX260140R020 ALX260140R100
ENZOALX260140R020EA 275 EUR
ENZOALX260140R020 ENZOALX260140R100
Z-WEHD-FMK
Proteine und Peptide
Inhibitor of caspase-1, -4 and -5
Irreversible, cell permeable inhibitor of caspase-1 and -5. Inhibits also caspase-4. Designed as a methyl ester to facilitate cell permeability.