BSA standard for protein assay
Lieferant: Uptima
UP36859A UP36859D
UPTIUP36859AEA 0 EUR
UPTIUP36859A UPTIUP36859D
BSA standard for protein assay
Tests Proteintests
High quality, optimised reagents for the determination of protein concentration by the bicinchoninic acid method.
  • High sensitivity and wide linear dynamic range