Anti-AMACR Rabbit Monoclonal Antibody [clone: 13H4]

Lieferant: IMMUNOLOGIC
ILM0422-C01 ILM0422-C05 ILM0422-C1
ILM0422-C01EA 113 EUR
ILM0422-C01 ILM0422-C05 ILM0422-C1
Anti-AMACR Rabbit Monoclonal Antibody [clone: 13H4]
Antikörper
Type: Primary
Antigen: AMACR
Clonality: Monoclonal
Clone: 13H4
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR