Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 14]

Lieferant: IMMUNOLOGIC
ILM7324-C01 ILM7324-C05 ILM7324-C1
ILM7324-C01EA 92.5 EUR
ILM7324-C01 ILM7324-C05 ILM7324-C1
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 14]
Antikörper
Type: Primary
Antigen: CTNNB1
Clonality: Monoclonal
Clone: 14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Chicken, Dog, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR