Anti-C4D Mouse Monoclonal Antibody [clone: 10-11]

Lieferant: IMMUNOLOGIC
ILM800416-C01 ILM800416-C05 ILM800416-C1
ILM800416-C01EA 412 EUR
ILM800416-C01 ILM800416-C05 ILM800416-C1
Anti-C4D Mouse Monoclonal Antibody [clone: 10-11]
Antikörper
Type: Primary
Antigen: C4D
Clonality: Monoclonal
Clone: 10-11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR