Anti-CD19 Mouse Monoclonal Antibody [clone: MX016]

Lieferant: IMMUNOLOGIC
ILM0705-C01 ILM0705-C05 ILM0705-C1
ILM0705-C01EA 182 EUR
ILM0705-C01 ILM0705-C05 ILM0705-C1
Anti-CD19 Mouse Monoclonal Antibody [clone: MX016]
Antikörper
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: MX016
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR