Anti-CD44 Mouse Monoclonal Antibody [clone: 156-3C11]

Lieferant: IMMUNOLOGIC
ILM9600-C1 ILM9600-C05 ILM9600-C01
ILM9600-C1EA 495 EUR
ILM9600-C1 ILM9600-C05 ILM9600-C01
Anti-CD44 Mouse Monoclonal Antibody [clone: 156-3C11]
Antikörper
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 156-3C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Baboon, Green Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR