Anti-PTPRC Mouse Monoclonal Antibody [clone: PD7/26+2B11]

Lieferant: IMMUNOLOGIC
ILM30511-C01 ILM30511-C05 ILM30511-C1
ILM30511-C01EA 68.7 EUR
ILM30511-C01 ILM30511-C05 ILM30511-C1
Anti-PTPRC Mouse Monoclonal Antibody [clone: PD7/26+2B11]
Antikörper
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: PD7/26+2B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR