Anti-PTPRC Mouse Monoclonal Antibody [clone: UCHL-1]

Lieferant: IMMUNOLOGIC
ILM07211-R10 ILM07211-R25
VWRKILM07211-R10EA 0 EUR
VWRKILM07211-R10 VWRKILM07211-R25
Anti-PTPRC Mouse Monoclonal Antibody [clone: UCHL-1]
Antikörper
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: UCHL-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Chimpanzee, Common Marmoset
Bestellen

TECHNISCHE DATEN

Antigen-Symbol PTPRC
Environmentally Preferable
Antigen-Name Protein tyrosine phosphatase/receptor type C
Antigen-Synonyme CD45|GP180|T200|LCA
Antikörper-Typ Primär
Klonalität Monoklonal
Konjugation Unconjugated
Klon UCHL-1
Reaktivität Common marmoset,Chimpanzee,Human
Wirt Mouse
Gen-ID 5788
Isotyp IgG2a kappa
ImmunoChemie Yes
Form Ready to use
Lagertemperatur 2°C - 8°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR