Anti-CD99 Rabbit Monoclonal Antibody [clone: EPR30974]

Lieferant: IMMUNOLOGIC
ILM1993-C01 ILM1993-C05 ILM1993-C1
ILM1993-C01EA 149 EUR
ILM1993-C01 ILM1993-C05 ILM1993-C1
Anti-CD99 Rabbit Monoclonal Antibody [clone: EPR30974]
Antikörper
Type: Primary
Antigen: CD99
Clonality: Monoclonal
Clone: EPR30974
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR