Anti-KRT18 Mouse Monoclonal Antibody [clone: DC10]

Lieferant: IMMUNOLOGIC
ILM4422-C01 ILM4422-C05 ILM4422-C1
ILM4422-C01EA 145 EUR
ILM4422-C01 ILM4422-C05 ILM4422-C1
Anti-KRT18 Mouse Monoclonal Antibody [clone: DC10]
Antikörper
Type: Primary
Antigen: KRT18
Clonality: Monoclonal
Clone: DC10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

TECHNISCHE DATEN

Antigen-Symbol KRT18
Environmentally Preferable
Antigen-Name Keratin 18
Antikörper-Typ Primär
Klonalität Monoklonal
Konjugation Unconjugated
Klon DC10
Reaktivität Human
Wirt Mouse
Gen-ID 3875
Isotyp IgG1
ELISA Yes
ImmunoChemie Yes
Immunpräzipitation Yes
Durchflusszytometrie Yes
Form Concentrated
Lagertemperatur 2°C - 8°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR