Anti-KRT19 Mouse Monoclonal Antibody [clone: BA17]

Lieferant: IMMUNOLOGIC
ILM1125-C01 ILM1125-C05 ILM1125-C1
ILM1125-C01EA 103 EUR
ILM1125-C01 ILM1125-C05 ILM1125-C1
Anti-KRT19 Mouse Monoclonal Antibody [clone: BA17]
Antikörper
Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: BA17
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR