Anti-DES Mouse Monoclonal Antibody [clone: D33]

Lieferant: IMMUNOLOGIC
ILM5116-C01 ILM5116-C05 ILM5116-C1
ILM5116-C01EA 75.6 EUR
ILM5116-C01 ILM5116-C05 ILM5116-C1
Anti-DES Mouse Monoclonal Antibody [clone: D33]
Antikörper
Type: Primary
Antigen: DES
Clonality: Monoclonal
Clone: D33
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR