Anti-EPCAM Mouse Monoclonal Antibody [clone: BER-EP4]

Lieferant: IMMUNOLOGIC
ILM2483-C01 ILM2483-C05 ILM2483-C1
ILM2483-C01EA 113 EUR
ILM2483-C01 ILM2483-C05 ILM2483-C1
Anti-EPCAM Mouse Monoclonal Antibody [clone: BER-EP4]
Antikörper
Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: BER-EP4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR