Anti-EPCAM Mouse Monoclonal Antibody [clone: MOC-31]

Lieferant: IMMUNOLOGIC
ILM4825-C01 ILM4825-C05 ILM4825-C1
ILM4825-C01EA 101 EUR
ILM4825-C01 ILM4825-C05 ILM4825-C1
Anti-EPCAM Mouse Monoclonal Antibody [clone: MOC-31]
Antikörper
Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: MOC-31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR