Anti-ESR1 Rabbit Monoclonal Antibody [clone: ZR2]

Lieferant: IMMUNOLOGIC
ILM2022-C01 ILM2022-C05 ILM2022-C1
ILM2022-C01EA 239 EUR
ILM2022-C01 ILM2022-C05 ILM2022-C1
Anti-ESR1 Rabbit Monoclonal Antibody [clone: ZR2]
Antikörper
Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: ZR2
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR