Anti-GFAP Mouse Monoclonal Antibody [clone: GA5]

Lieferant: IMMUNOLOGIC
ILM2670-C01 ILM2670-C05 ILM2670-C1
ILM2670-C01EA 61 EUR
ILM2670-C01 ILM2670-C05 ILM2670-C1
Anti-GFAP Mouse Monoclonal Antibody [clone: GA5]
Antikörper
Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: GA5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Cow, Pig, Rabbit, Chicken
Bestellen

TECHNISCHE DATEN

Antigen-Symbol GFAP GFAP
Environmentally Preferable
Antigen-Name Glial fibrillary acidic protein Glial fibrillary acidic protein
Antigen-Synonyme FLJ45472 FLJ45472
Antikörper-Typ Primär Primär
Klonalität Monoklonal Monoklonal
Konjugation Unconjugated Unconjugated
Klon GA5 GA5
Reaktivität Pig,Rabbit,Chicken,Human,Bovine,Rat,Mouse Pig,Rabbit,Chicken,Human,Bovine,Rat,Mouse
Wirt Mouse Mouse
Gen-ID 2670 2670
Isotyp IgG1 IgG1
Western blot Yes Yes
ImmunoChemie Yes Yes
ImmunFluoreszenz Yes Yes
Durchflusszytometrie Yes Yes
Immunogen GFAP isolated from pig spinal cord
Form Concentrated Concentrated
Molecular weight 50 kDa
Lagertemperatur 2°C - 8°C 2°C - 8°C
Validierte Anwendungen Immunohistochemical Staining of Formalin-Fixed Tissue Sections

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR