Anti-INHA Mouse Monoclonal Antibody [clone: R1]

Lieferant: IMMUNOLOGIC
ILM57716-C01 ILM57716-C05 ILM57716-C1
ILM57716-C01EA 227 EUR
ILM57716-C01 ILM57716-C05 ILM57716-C1
Anti-INHA Mouse Monoclonal Antibody [clone: R1]
Antikörper
Type: Primary
Antigen: INHA
Clonality: Monoclonal
Clone: R1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Sheep, Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR