Anti-PMEL Mouse Monoclonal Antibody [clone: HMB45]

Lieferant: IMMUNOLOGIC
ILM576211-C01 ILM576211-C05 ILM576211-C1
ILM576211-C01EA 78.8 EUR
ILM576211-C01 ILM576211-C05 ILM576211-C1
Anti-PMEL Mouse Monoclonal Antibody [clone: HMB45]
Antikörper
Type: Primary
Antigen: PMEL
Clonality: Monoclonal
Clone: HMB45
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR