Anti-MLH1 Mouse Monoclonal Antibody [clone: G168-728]

Lieferant: IMMUNOLOGIC
ILM8704-C01 ILM8704-C05 ILM8704-C1
ILM8704-C01EA 166 EUR
ILM8704-C01 ILM8704-C05 ILM8704-C1
Anti-MLH1 Mouse Monoclonal Antibody [clone: G168-728]
Antikörper
Type: Primary
Antigen: MLH1
Clonality: Monoclonal
Clone: G168-728
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

TECHNISCHE DATEN

Antigen-Symbol MLH1
Environmentally Preferable
Antigen-Name MutL homolog 1
Antigen-Synonyme COCA2|HNPCC|FCC2|HNPCC2
Antikörper-Typ Primär
Klonalität Monoklonal
Konjugation Unconjugated
Klon G168-728
Reaktivität Human
Wirt Mouse
Gen-ID 4292
Isotyp IgG1
ImmunoChemie Yes
Form Concentrated
Lagertemperatur 2°C - 8°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR