Anti-NAPSA Mouse Monoclonal Antibody [clone: MX0165]

Lieferant: IMMUNOLOGIC
ILM0704-C01 ILM0704-C05 ILM0704-C1
ILM0704-C01EA 248 EUR
ILM0704-C01 ILM0704-C05 ILM0704-C1
Anti-NAPSA Mouse Monoclonal Antibody [clone: MX0165]
Antikörper
Type: Primary
Antigen: NAPSA
Clonality: Monoclonal
Clone: MX0165
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR