Anti-RABEPK Rabbit Monoclonal Antibody [clone: ZR8]

Lieferant: IMMUNOLOGIC
ILM0022-C01 ILM0022-C05 ILM0022-C1
ILM0022-C01EA 224 EUR
ILM0022-C01 ILM0022-C05 ILM0022-C1
Anti-RABEPK Rabbit Monoclonal Antibody [clone: ZR8]
Antikörper
Type: Primary
Antigen: RABEPK
Clonality: Monoclonal
Clone: ZR8
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR