Anti-TP53 Mouse Monoclonal Antibody [clone: DO7]

Lieferant: IMMUNOLOGIC
ILM27011-C01 ILM27011-C05 ILM27011-C1
ILM27011-C01EA 78.8 EUR
ILM27011-C01 ILM27011-C05 ILM27011-C1
Anti-TP53 Mouse Monoclonal Antibody [clone: DO7]
Antikörper
Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: DO7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Monkey, Cow
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR