Anti-PAX8 Mouse Monoclonal Antibody [clone: PAX8/1492]

Lieferant: IMMUNOLOGIC
ILM7849-C1 ILM7849-C01 ILM7849-C05
ILM7849-C1EA 1060 EUR
ILM7849-C1 ILM7849-C01 ILM7849-C05
Anti-PAX8 Mouse Monoclonal Antibody [clone: PAX8/1492]
Antikörper
Type: Primary
Antigen: PAX8
Clonality: Monoclonal
Clone: PAX8/1492
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR