Anti-S100B Rabbit Polyclonal Antibody

Lieferant: IMMUNOLOGIC
ILP2025-C01 ILP2025-C05 ILP2025-C1
ILP2025-C01EA 102 EUR
ILP2025-C01 ILP2025-C05 VWRKILP2025-C1
Anti-S100B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: S100B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR