Anti-SMAD4 Mouse Monoclonal Antibody [clone: B-8]

Lieferant: IMMUNOLOGIC
ILM9124-C01 ILM9124-C05 ILM9124-C1
ILM9124-C01EA 171 EUR
ILM9124-C01 ILM9124-C05 ILM9124-C1
Anti-SMAD4 Mouse Monoclonal Antibody [clone: B-8]
Antikörper
Type: Primary
Antigen: SMAD4
Clonality: Monoclonal
Clone: B-8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR