Anti-SMTN Mouse Monoclonal Antibody [clone: R4A]

Lieferant: IMMUNOLOGIC
ILM1206-C01 ILM1206-C05 ILM1206-C1
ILM1206-C01EA 189 EUR
ILM1206-C01 ILM1206-C05 ILM1206-C1
Anti-SMTN Mouse Monoclonal Antibody [clone: R4A]
Antikörper
Type: Primary
Antigen: SMTN
Clonality: Monoclonal
Clone: R4A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR