Anti-SOX10 Rabbit Monoclonal Antibody

Lieferant: IMMUNOLOGIC
ILP3833-C01 ILP3833-C05 ILP3833-C1 ILP3833-R7
ILP3833-C01EA 223 EUR
ILP3833-C01 ILP3833-C05 VWRKILP3833-C1 VWRKILP3833-R7
Anti-SOX10 Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SOX10
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR