Anti-WT1 Mouse Monoclonal Antibody [clone: WT49]

Lieferant: IMMUNOLOGIC
ILM3951-C01 ILM3951-C05 ILM3951-C1
ILM3951-C01EA 145 EUR
ILM3951-C01 ILM3951-C05 ILM3951-C1
Anti-WT1 Mouse Monoclonal Antibody [clone: WT49]
Antikörper
Type: Primary
Antigen: WT1
Clonality: Monoclonal
Clone: WT49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR