Anti-ZAP70 Mouse Monoclonal Antibody [clone: 2F3.2]

Lieferant: IMMUNOLOGIC
ILM7535-C01 ILM7535-C05 ILM7535-C1
ILM7535-C01EA 112 EUR
ILM7535-C01 ILM7535-C05 ILM7535-C1
Anti-ZAP70 Mouse Monoclonal Antibody [clone: 2F3.2]
Antikörper
Type: Primary
Antigen: ZAP70
Clonality: Monoclonal
Clone: 2F3.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR