Anti-IL-22BP Rabbit Monoclonal Antibody [clone: 28]

Lieferant: Sino Biological
11025-R028-50 11025-R028-100
SINO11025-R028-50EA 0 EUR
SINO11025-R028-50 SINO11025-R028-100
Anti-IL-22BP Rabbit Monoclonal Antibody [clone: 28]
Antikörper
Anti-IL-22BP antibody

Type: Primary
Antigen: IL-22BP
Clonality: Monoclonal
Clone: Clone: 28
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR