Anti-IgG Goat Polyclonal Antibody (iFluor® 568)

Lieferant: AAT BIOQUEST
16622 16804
AATB16622EA 120 EUR
AATB16622 AATB16804
Anti-IgG Goat Polyclonal Antibody (iFluor® 568)
Antikörper
iFluor® 568 is a bright red fluorescent dye.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins

Bitte beachten For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: iFluor™ 568
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR