Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (APC/iFluor® 700)

Lieferant: AAT BIOQUEST
100211F0 100211F1 100211F2
AATB100211F0EA 319 EUR
AATB100211F0 AATB100211F1 AATB100211F2
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (APC/iFluor® 700)
Antikörper
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: APC/iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR